Learn More
DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
Brand:  DWK Life Sciences 223684
68.34 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10747944
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Compliance
Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.27 in.
Clear
Borosilicate Glass
For lyophilization, packaging, sampling and freeze drying applications
Autoclavable
38 mm
7 mm
17 mm
3 mL
0.8 dr.
Crimp Cap
ASTM E438, USP Type I, Type I, Class A