missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Straight Plug Style Bromobutyl rubber Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Brand: DWK Life Sciences 224100-331
This item is not returnable.
View return policy
Description
- Available in choice of different materials (silicone and bromobutyl)
Not recommended for vials with a blow-back feature.
Specifications
3cm | |
50 Durometer | |
Gray | |
Straight Plug Stopper |
Fits 1.5cm I.D., 3cm O.D. mouth of vials and serum bottles | |
Autoclavable | |
Bromobutyl Rubber |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Straight Plug Style Bromobutyl rubber Stoppers for Vials and Serum Bottles > Gray bromobutyl; Mouth I.D. x O. D.: 15 x 30mm
Spot an opportunity for improvement?Share a Content Correction