missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

£24.20 - £42.15

Specifications

Color White
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner Polyethylene Foam
View More Specs

Products 9
Product Code Brand Neck Finish Material No. per Case Price Quantity & Availability  
Product Code Brand Neck Finish Material No. per Case Price Quantity & Availability  
11798899
View Documents
DWK Life Sciences
239286
43-400 Polypropylene 72 Cs.
£25.15
Pack of 72
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11738909
View Documents
DWK Life Sciences
239290
58-400 Polypropylene 72 Cs.
£40.90
Pack of 72
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
 
11718909
View Documents
DWK Life Sciences
239288
48-400 Polypropylene 72 Cs.
£34.40
Pack of 72
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11708899
View Documents
DWK Life Sciences
239273
13-425 Polypropylene 144 Cs.
£24.55
Pack of 144
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11728909
View Documents
DWK Life Sciences
239289
53-400 Glass 72 Cs.
£40.00
Pack of 72
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11758899
View Documents
DWK Life Sciences
239281
24-400 Polypropylene 144 Cs.
£34.40
Pack of 144
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11758909
View Documents
DWK Life Sciences
239292
70-400 Polypropylene 48 Cs.
£42.15
Pack of 48
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11768899
View Documents
DWK Life Sciences
239283
28-400 Polypropylene 144 Cs.
£28.80
Pack of 144
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11788899
View Documents
DWK Life Sciences
239285
38-400 Polypropylene 72 Cs.
£24.20
Pack of 72
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
Description

Description

  • Liner provides tight seal
  • Liner Material: Foam, Polyethylene
  • Polypropylene
  • Screw Cap
  • White

Use these caps with glass bottles or vials

Order Info

Each pack contains 72

Specifications

Specifications

White
ISO 9001
Glass bottle or vial
Polyethylene Foam
Videos
SDS
Documents

Documents

Product Certifications