missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ White 96-Well Immuno Plates
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Brand: Thermo Scientific™ 7705
124.18 GBP valid until 2024-12-31
Use promo code "21894" to get your promotional price.
This item is not returnable.
View return policy
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Choice of three surface types for optimal binding
- Certified binding homogeneity/reproducibility
Choice of Surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Specifications
Hydrophobic | |
Chemiluminescence | |
50 Cases | |
White | |
Polystyrene | |
96 | |
Microfluor 1 | |
Fluorescence | |
10 | |
Lipids |
Standard | |
8 x 12 | |
Adsorption | |
Microplate | |
330 μL | |
280 μL | |
Round, Flat Bottom (F-well) | |
50 | |
None |