Learn More
Thermo Scientific™ White 96-Well Immuno Plates
GREENER_CHOICE
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Brand:  Thermo Scientific™ 7905
174.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11952984
  • £214.00 / Pack of 50
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility
Choice of Surface:
  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Slightly Hydrophilic
With Lid
8 x 12
Adsorption
Microplate
2280 μL
280 μL
Round, Flat Bottom (F-well)
50
None
Standard
Chemiluminescence
50 Cases
White
Polystyrene
96
Microfluor 2
Fluorescence
10
Antibodies (Glycoproteins) and Proteins