missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Plastic General Purpose Trays

Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12367237

  • £85.97 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications

Specifications

570 x 540mm
570 mm
Tray
Safe containment of spillages, lab bench protection
1 Each
White
Plastic
540 mm
Liner
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers