Learn More
Fisherbrand™ White Plastic General Purpose Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
Brand:  Fisherbrand™ CSR-TW6
 View more versions of this product
Product Code. 12397237
  • £134.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Specifications
1130 x 540mm
1130 mm
Tray
Safe containment of spillages, lab bench protection
1 Each
White
Plastic
540 mm
Liner
Rectangular
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products