Learn More
Gosselin™ White Specimen Containers with Red Cap
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand:  Gosselin™ TP52C-023
Code : NEW
Additional Details : Weight : 8.05500kg
 View more versions of this product
Product Code. 15458834
  • £150.23 / Pack of 380
Estimated Shipment: 25-10-2024
to see stock.
Description
Description
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
Specifications
Specifications
Specimen Container
Polypropylene
Sterile
Red
White
380/Cs.
Yes
Straight
125 mL
Screw Cap
No Label
380/Pk.