missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Wide Mouth Round Bottle, HDPE, PP/Foamed PE Lined Lid
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
3840 mL
Description
- High Density Polyethylene (HDPE) in a natural color
- Provides good chemical resistance
- Without caps or supplied with pre-attached white polypropylene screw caps with PE liners

Specifications
Specifications
Capacity (Metric) | 3840 mL |
Color | Natural |
Material | HDPE |
Quantity | 4/Case |
Closure Color | White |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Product Type | Storage Jar |
Closure Liner | Polyethylene Foam |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Wide Mouth Round Bottle, HDPE, PP/Foamed PE Lined Lid >
Spot an opportunity for improvement?Share a Content Correction