missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Wide Neck Erlenmeyer Flask

Wide Neck Erlenmeyer Flask features a beaded rim and graduation marks.

Brand:  BRAND™ 92863

377.00 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.



Additional Details : Weight : 55.25000kg

 View more versions of this product

Product Code. 10440293

  • £402.00 / Pack of 10
Estimated Shipment: 11-10-2024
to see stock.
Add to basket

Description

Description

  • Borosilicate Glass 3.3
  • DIN ISO 24450
  • With beaded rim and graduation
Specifications

Specifications

2000 mL
Wide
Beaded
Borosilicate Glass (3.3)
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers