missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations

Specifications
Specifications
Boiling Point | 118°C |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000%, <2.0000% |
Quantity | 6 x 1 L |
Linear Formula | ICl |
MDL Number | MFCD00011354 |
UN Number | UN2789 |
Beilstein | 3587194 |
Flash Point | 40°C |
Synonym | Iodine monochloride solution, Iodine according to Wijs for the iodine value determination, Chloroiodide solution, Wijs solution |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling



Product Identifier
- Wijs solution
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Acetic acid, iodinemonochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction