missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ William's E Medium, no glutamine
William's E Medium, no glutamine
£29.55 - £258.00
Specifications
Form | Liquid |
---|---|
Product Type | William's E Medium |
Without Additives | No Glutamine, No HEPES |
Product Code | Brand | Quantity | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | For Use With (Application) | Price | Quantity & Availability | ||||
12599059
|
Gibco™
12551032 |
500 mL | Hepatocytes and hepatic models, mammalian cell culture, pharma and biopharma, primary cell culture |
£37.76
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11570606
|
Gibco™
22551022 |
500 mL | Mammalian cell culture |
£29.55
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11580606
|
Gibco™
22551089 |
10 x 500 mL | Mammalian cell culture |
£258.00
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ William's E Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ William's E Medium product made in our Grand Island facility (12551-032). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in your specific region, it should be used for laboratory use only.
Specifications
Liquid | |
William's E Medium | |
No Glutamine, No HEPES |