missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Wilmad-LabGlass™ High-Throughput NMR Tubes
Double the writing area allows for easy sample marking and identification.
£160.16 - £280.80
Specifications
Diameter (Metric) Outer | 5±0.02 mm |
---|---|
Length (English) | 7 in. |
Thickness (Metric) Wall | 0.43 mm |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
16695672
|
SP
66999-5751 |
50 Pack |
£160.16
Pack of 50 |
In Stock
Log in to see stock. |
||||||
16685672
|
SP
66999-5702 |
100 Pack |
£280.80
Pack of 100 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Average camber of 60 microns ensures spectrum clarity for small molecule (MW <250) samples up to 600 MHz.
- 2x larger marking area eliminates cramped and unreadable sample names
- Silkscreened writing area keeps sample names from easily wiping off
- Designed for routine use in most low to mid field NMR spectrometers
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimensions
Note: For experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications
5±0.02 mm | |
7 in. | |
0.43 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SP Wilmad-LabGlass™ High-Throughput NMR Tubes