Learn More
Cole Parmer™ Write-On Tape
Use to label/identify laboratory items/products.
Brand: Cole Parmer™ 06530-21
This item is not returnable.
View return policy
Description
- Identify with writing and color.
- Durable tape is waterproof, and oil and acid resistant.
- Can be used with pencil, pen, or permanent marker.
- Temperature resistant.
Specifications
White | |
6/Pk. | |
14 yd. x 0.5 in. | |
0.5 in. |
6/Pk. | |
-23°C to +121°C | |
14 yd. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Cole Parmer™ Write-On Tape > White, 1/2 in. x 14 yds, 6 PK
Spot an opportunity for improvement?Share a Content Correction