Learn More
Kimberly-Clark™ WypAll™ L40 Fabric Wipers
GREENER_CHOICE
Use these wipers for industrial tasks such as cleaning, process line wiping and soaking up large spills. WypAll™ L40 Paper Wipers are made from a specialized fabric, featuring double re-creped (DRC) base sheet technology for balance of softness and strength.
£321.00 - £337.00
Specifications
Product Type Medium-duty Industrial Wiper
Material Double Re-crepe Cellulose
Color White
For Use With (Application) A limited life wiper using double re-creped (DRC) base sheet technology, layered fabric that’s strong, thick and highly absorbent so it cleans up fast without falling apart, does the job with fewer wipers and helps reduce costs
Ply 1-ply
View More Specs
Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
11778038
GREENER_CHOICE
View Documents
Kimberly-Clark™
7462
25 x 41 cm (9.8 x 16.1 in.)
£337.00
Pack of 9
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
10627645
GREENER_CHOICE
View Documents
Kimberly-Clark™
7471
12 x 12.5 in.
£321.00
Pack of 18
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Large, perforated roll for use in busy areas
  • Can be used with a dispensing system to help control use and reduce waste
Specifications
Specifications
Medium-duty Industrial Wiper
White
1-ply
Double Re-crepe Cellulose
A limited life wiper using double re-creped (DRC) base sheet technology, layered fabric that’s strong, thick and highly absorbent so it cleans up fast without falling apart, does the job with fewer wipers and helps reduce costs
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Kimberly-Clark™ WypAll™ L40 Fabric Wipers