Learn More
ABEBA™ X-Light Black Occupational Shoes O2 ESD
Black X-light occupational shoes with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Brand:  ABEBA™ 7131138/38
Additional Details : Weight : 0.73000kg
 View more versions of this product
Product Code. 17151555
  • £84.98 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Lightweight, comfortable occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Black smooth leather
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Specifications
Black
38
Unisex
Water Resistant
Leather
Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Low-cut
O2 SRC ESD