Learn More
ABEBA™ X-Light Black Safety Slip-On Shoes S2
ABEBA™ X-Light Black Safety Slip-On Shoes S2 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
Brand:  ABEBA™ 7131029/45
53.65 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 16398281
  • £92.10 / 1 pair
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mon PU outsole SRC
  • Good cushioning
  • Black washable microfiber
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Specifications
CE EN ISO 20345:2012 S2, SRC, ESD
Breathable lining with Silver ions
45
Polyurethane
Steel
CE EN ISO 20345:2012, DIN EN 61340
Black
Microfiber
Slip and abrasion resistant, shock absorbing, ESD dissipative
Slip-on shoes
1 Pair
Water