Learn More
ABEBA™ X-Light Black/Yellow Safety Trainers S3 ESD
Black/yellow X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
Brand: ABEBA™ 7131860/41
Additional Details : Weight : 0.86000kg
Description
- Lightweight, comfortable occupational shoes
- Comply to EN ISO 20345: 2012, S3 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant Mono PU outsole SRC
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Black and yellow Cordura
- Silver ions lining
- Replaceable active comfort insole
Specifications
Black, Yellow | |
41 | |
Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340 | |
Trainers | |
S3 SRC ESD |
Cordura | |
Steel | |
Unisex | |
Water Resistant |