Learn More
ABEBA™ X-Light White Occupational Shoes O2 ESD
White X-light occupational shoes, with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Brand: ABEBA™ 7131124/46
Additional Details : Weight : 0.76000kg
Description
- Lightweight, comfortable white occupational shoes
- Comply to EN ISO 20347: 2012, O2 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant Mono PU outsole SRC
- White technical fabric
- Silver ions lining
- Replaceable active comfort insole
Specifications
White | |
46 | |
Unisex | |
Water Resistant |
Synthetic fabric | |
Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340 | |
Low-cut | |
O2 SRC ESD |