missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH Amide XP Method Validation Kit

This method validation kit consists of three batches of XBridge BEH Amide columns packed with three different batches of sorbent.

Brand:  Waters Corporation 186006229

Encompass

 View more versions of this product

Product Code. 17173612

  • £2805.00 / Pack of 3
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes three batches of XBridge BEH Amide columns packed with three different batches of sorbent.
Specifications

Specifications

3 mm
18000 psi (1240 Bar)
2.5 μm
3/Pk.
No
50 mm
Spherical
130 Å
L68
eXtended Performance (XP) Method Validation Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers