missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge Phenyl Column

XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.

Brand:  Waters Corporation 186003342

Encompass

 View more versions of this product

Product Code. 13135039

  • £877.50 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Specifications

Specifications

3 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
30 mm
Spherical
130 Å
L11
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers