Learn More
Waters Corporation XBridge Phenyl Column
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
£942.00 - £1218.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L11 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
13185039
![]() |
Waters Corporation
186003348 |
30 mm | 4.6 mm | 5 μm |
£942.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13175019
![]() |
Waters Corporation
186003322 |
50 mm | 2.1 mm | 3.5 μm |
£999.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13115029
![]() |
Waters Corporation
186003328 |
100 mm | 3 mm | 3.5 μm |
£1074.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10628277
![]() |
Waters Corporation
186003329 |
150 mm | 3 mm | 3.5 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13195029
![]() |
Waters Corporation
186003338 |
50 mm | 2.1 mm | 5 μm |
£964.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10033994
![]() |
Waters Corporation
186003349 |
50 mm | 4.6 mm | 5 μm |
£964.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10210515
![]() |
Waters Corporation
186003351 |
100 mm | 4.6 mm | 5 μm |
£1039.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13105049
![]() |
Waters Corporation
186003352 |
150 mm | 4.6 mm | 5 μm |
£1108.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13115049
![]() |
Waters Corporation
186003353 |
250 mm | 4.6 mm | 5 μm |
£1218.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13125019
![]() |
Waters Corporation
186003317 |
50 mm | 1 mm | 3.5 μm |
£999.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13145019
![]() |
Waters Corporation
186003319 |
150 mm | 1 mm | 3.5 μm |
£1149.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13165019
![]() |
Waters Corporation
186003321 |
30 mm | 2.1 mm | 3.5 μm |
£970.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10298006
![]() |
Waters Corporation
186003323 |
100 mm | 2.1 mm | 3.5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542877
![]() |
Waters Corporation
186003324 |
150 mm | 2.1 mm | 3.5 μm |
£1149.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13195019
![]() |
Waters Corporation
186003326 |
30 mm | 3 mm | 3.5 μm |
£970.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13105029
![]() |
Waters Corporation
186003327 |
50 mm | 3 mm | 3.5 μm |
£999.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13135029
![]() |
Waters Corporation
186003331 |
30 mm | 4.6 mm | 3.5 μm |
£970.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13145029
![]() |
Waters Corporation
186003332 |
50 mm | 4.6 mm | 3.5 μm |
£999.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13155029
![]() |
Waters Corporation
186003333 |
75 mm | 4.6 mm | 3.5 μm |
£1017.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13165029
![]() |
Waters Corporation
186003334 |
100 mm | 4.6 mm | 3.5 μm |
£1074.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13185029
![]() |
Waters Corporation
186003337 |
30 mm | 2.1 mm | 5 μm |
£942.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
12376223
![]() |
Waters Corporation
186003339 |
100 mm | 2.1 mm | 5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13115039
![]() |
Waters Corporation
186003340 |
150 mm | 2.1 mm | 5 μm |
£1108.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13135039
![]() |
Waters Corporation
186003342 |
30 mm | 3 mm | 5 μm |
£942.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13145039
![]() |
Waters Corporation
186003343 |
50 mm | 3 mm | 5 μm |
£964.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13155039
![]() |
Waters Corporation
186003344 |
100 mm | 3 mm | 5 μm |
£1039.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10034783
![]() |
Waters Corporation
186003345 |
150 mm | 3 mm | 5 μm |
£1108.50
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13165039
![]() |
Waters Corporation
186003346 |
250 mm | 3 mm | 5 μm |
£1218.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
13195039
![]() |
Waters Corporation
186003350 |
75 mm | 4.6 mm | 5 μm |
£999.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
Description
- This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Specifications
6000 psi (415 Bar) | |
130 Å | |
L11 | |
Column |
Spherical | |
1/Ea. | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XBridge Phenyl Column