missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge Prep C8 Column

The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent.

Brand:  Waters Corporation 186003083

Encompass

 View more versions of this product

Product Code. 13243010

  • £8610.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance.
  • The trifunctionally bonded sorbent offers you the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
Specifications

Specifications

30 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
150 mm
Spherical
130 Å
L7
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers