Learn More
Waters Corporation XBridge Prep C8 Column
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent.
£339.00 - £20700.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L7 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
13262940
|
Waters Corporation
186002997 |
100 mm | 30 mm | 6 μm |
£6870.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13232940
|
Waters Corporation
186002994 |
100 mm | 19 mm | 5 μm |
£3780.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13242940
|
Waters Corporation
186002995 |
150 mm | 19 mm | 5 μm |
£4515.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13243010
|
Waters Corporation
186003083 |
150 mm | 30 mm | 5 μm |
£8610.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13184989
|
Waters Corporation
186003269 |
75 mm | 30 mm | 5 μm |
£5925.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13253260
|
Waters Corporation
186003934 |
50 mm | 50 mm | 5 μm |
£8715.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13293260
|
Waters Corporation
186003938 |
100 mm | 50 mm | 5 μm |
£11910.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13222940
|
Waters Corporation
186002993 |
50 mm | 19 mm | 5 μm |
£2265.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13252940
|
Waters Corporation
186002996 |
50 mm | 30 mm | 5 μm |
£4650.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13283310
|
Waters Corporation
186004003 |
10 mm | 10 mm | 10 μm |
£339.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
10438086
|
Waters Corporation
186004006 |
10 mm | 19 mm | 10 μm |
£573.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13223320
|
Waters Corporation
186004007 |
50 mm | 19 mm | 10 μm |
£2175.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13233320
|
Waters Corporation
186004008 |
100 mm | 19 mm | 10 μm |
£3600.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13243320
|
Waters Corporation
186004009 |
150 mm | 19 mm | 10 μm |
£4275.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
10477476
|
Waters Corporation
186004010 |
250 mm | 19 mm | 10 μm |
£5415.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13263320
|
Waters Corporation
186004011 |
150 mm | 30 mm | 10 μm |
£9690.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13273320
|
Waters Corporation
186004012 |
250 mm | 30 mm | 10 μm |
£15150.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13283320
|
Waters Corporation
186004013 |
50 mm | 50 mm | 10 μm |
£8745.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13293320
|
Waters Corporation
186004014 |
100 mm | 50 mm | 10 μm |
£11910.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13203330
|
Waters Corporation
186004015 |
150 mm | 50 mm | 10 μm |
£16200.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13213330
|
Waters Corporation
186004016 |
250 mm | 50 mm | 10 μm |
£20700.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
13243330
|
Waters Corporation
186004023 |
250 mm | 19 mm | 5 μm |
£5805.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
Beschreibung
- Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance.
- The trifunctionally bonded sorbent offers you the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
Spezifikation
6000 psi (415 Bar) | |
130 Å | |
L7 | |
Column |
Spherical | |
1/Ea. | |
Yes |