Learn More
TNCBio™ XerumFree™
Defined and ACF additive for serum-free cell culture.
Brand: TNCBio™ XF212-0500-12
4531.50 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 6.00000kg
Description
Defined additive for serum-free cell culture. Animal/human compound free.
Specifications
Transparent liquid | |
Liquid | |
Sterile-filtered | |
285 - 325 mOsm/kg | |
12 x 500 mL |
4°C | |
Cell Culture Supplement | |
Sterile-filtered | |
7 to 8 |