missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNCBio™ XerumFree™
Defined and ACF additive for serum-free cell culture.
£120.00 - £4770.00
Specifications
Content And Storage | 4°C |
---|---|
Form | Liquid |
Product Type | Cell Culture Supplement |
Sterility | Sterile-filtered |
Sterilization Method | Sterile-filtered |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17896927
|
TNCBio™
XF212-0100 |
100 mL |
£120.00
Each |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17806937
|
TNCBio™
XF212-0500 |
500 mL |
£433.00
Each |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17886927
|
TNCBio™
XF212-0500-12 |
12 x 500 mL |
£4770.00
Pack of 12 |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Defined additive for serum-free cell culture. Animal/human compound free.
Specifications
4°C | |
Cell Culture Supplement | |
Sterile-filtered | |
7 to 8 |
Liquid | |
Sterile-filtered | |
285 - 325 mOsm/kg |