Learn More
Invitrogen™ XIAP Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHXIAPX5
Description
The Human XIAP ELISA quantitates Hu XIAP in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu XIAP. Principle of the method The Human XIAP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.Specifications
P98170 | |
250 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
331 | |
<12% | |
Standard, Chromogen, Stop Solution, Wash Buffer, Adhesive Plate Covers, Assay Diluent Concentrate, SAV-HRP, Biotinylated Detection Antibody, Pre-coated 96 well plates | |
5 x 96 Tests | |
Apoptosis | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.46 ng/mL to 600 ng/mL | |
0.25 ng/mL to 600 ng/mL | |
Biotin | |
Other Proteins | |
This ELISA antibody pair can recognize human and mouse XIAP. | |
Human | |
Colorimetric Microplate Reader | |
API3,BIRC4,IAP-3,ILP1,MIHA,XLP2,hIAP-3,hIAP3 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
RP1-315G1.5, API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3 | |
1 hr. 20 min. |