missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XIAP Human ELISA Kit

Sandwich ELISA Kit

£546.00 - £4280.00

Specifications

Accession Number P98170
Assay Sensitivity 250 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405183
View Documents
Invitrogen™
EHXIAP
96 Tests
£546.00
1 set
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
17814230
View Documents
Invitrogen™
EHXIAPX10
10 x 96 Tests
£4280.00
Pack of 10
Estimated Shipment: 20-12-2024
to see stock.
Add to basket
 
17115609
View Documents
Invitrogen™
EHXIAPX5
5 x 96 Tests
£2320.00
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
Description

Description

The Human XIAP ELISA quantitates Hu XIAP in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu XIAP. Principle of the method The Human XIAP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.
Specifications

Specifications

P98170
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
API3,BIRC4,IAP-3,ILP1,MIHA,XLP2,hIAP-3,hIAP3
<10%
RUO
2°C to 8°C
Human
4 hr. 45 min.
250 pg/mL
Biotin
Other Proteins
This ELISA antibody pair can recognize human and mouse XIAP.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
331
<12%
HRP
Apoptosis
RP1-315G1.5, API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3
1 hr. 20 min.
Videos
SDS
Documents

Documents

Product Certifications