Learn More
Cytiva XK Chromatography Columns (Empty)
Ensure trouble-free operation for low to medium pressure. Cytiva XK Chromatography Columns (Empty) connect directly to ÄKTA design instruments or other liquid chromatography systems. The user-friendly ensures reproducible, accurate packing.
£494.00 - £1209.00
Specifications
Product Type | XK Chromatography Columns (Empty) |
---|---|
Material | Borosilicate glass |
Particle Size | 10μm pore size |
Support Type | Polypropylene (reinforced glass fiber)/Polyamide Net |
Column Type | Empty Chromatography Column |
Product Code | Brand | Height (Metric) | Diameter (Metric) Inner | Bed Volume | Flow Rate | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Diameter (Metric) Inner | Bed Volume | Flow Rate | Price | Quantity & Availability | ||
11320342
|
Cytiva
28-9889-53 |
300mm | 50mm | Bed height (one adapter): 140 to 280mm Bed height (two adapters): 0 to 280mm |
Inlet/Outlet (i.d.): 1.55mm |
£1013.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11330332
|
Cytiva
28-9889-37 |
200mm | 16mm | Bed height (one adapter): 25 to 155mm Bed height (two adapters): 0 to 155mm |
Inlet/Outlet (i.d.): 0.75mm |
£494.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||
11370332
|
Cytiva
28-9889-48 |
200mm | 26mm | Bed height (one adapter): 10 to 125mm Bed height (two adapters): 0 to 125mm |
Inlet/Outlet (i.d.): 0.75mm |
£558.00
Each |
Estimated Shipment: 10-10-2024
Log in to see stock. |
||
11380332
|
Cytiva
28-9889-49 |
400mm | 26mm | Bed height (one adapter): 230 to 350mm Bed height (two adapters): 85 to 350mm |
Inlet/Outlet (i.d.): 0.75mm |
£649.00
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
11338410
|
Cytiva
28-9889-52 |
200mm | 50mm | Bed height (one adapter): 0 to 140mm Bed height (two adapters): 0 to 140mm |
Inlet/Outlet (i.d.): 1.55mm |
£840.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11360332
|
Cytiva
28-9889-47 |
1000mm | 16mm | Bed height (one adapter): 825 to 950mm Bed height (two adapters): 680 to 950mm |
Inlet/Outlet (i.d.): 0.75mm |
£739.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11340332
|
Cytiva
28-9889-38 |
400mm | 16mm | Bed height (one adapter): 225 to 350mm Bed height (two adapters): 80 to 350mm |
Inlet/Outlet (i.d.): 0.75mm |
£597.00
Each |
Estimated Shipment: 10-10-2024
Log in to see stock. |
||
11350332
|
Cytiva
28-9889-46 |
700mm | 16mm | Bed height (one adapter): 525 to 650mm Bed height (two adapters): 380 to 650mm |
Inlet/Outlet (i.d.): 0.75mm |
£672.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11390332
|
Cytiva
28-9889-50 |
700mm | 26mm | Bed height (one adapter): 530 to 650mm Bed height (two adapters): 385 to 650mm |
Inlet/Outlet (i.d.): 0.75mm |
£725.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11330342
|
Cytiva
28-9889-64 |
600mm | 50mm | Bed height (one adapter): 400 to 560mm Bed height (two adapters): 260 to 560mm |
Inlet/Outlet (i.d.): 1.55mm |
£1068.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11300342
|
Cytiva
28-9889-51 |
1000mm | 26mm | Bed height (one adapter): 830 to 950mm Bed height (two adapters): 685 to 950mm |
Inlet/Outlet (i.d.): 0.75mm |
£835.00
Each |
Estimated Shipment: 10-10-2024
Log in to see stock. |
||
11340342
|
Cytiva
28-9889-65 |
1000mm | 50mm | Bed height (one adapter): 810 to 950mm Bed height (two adapters): 650 to 950mm |
Inlet/Outlet (i.d.): 1.55mm |
£1209.00
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||
Description
XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.
Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.
- Plunger design to ensure a homogeneous buffer distribution and reproducible packing
- Adapter QuickLock mechanism that facilitates column handling and cleaning
- Fittings for direct connection to chromatographiy systems
- Cooling jacket for sensitive purifications
- Extensive range of spare parts available, facilitating maintenance
Specifications
XK Chromatography Columns (Empty) | |
10μm pore size | |
Empty Chromatography Column |
Borosilicate glass | |
Polypropylene (reinforced glass fiber)/Polyamide Net | |
Liquid chromatography at low to medium pressure |
Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.
Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.