missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XSelect CSH Fluoro-Phenyl Column

Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases.

Brand:  Waters Corporation 186005331

Encompass

 View more versions of this product

Product Code. 17193042

  • £1035.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The non-endcapped sorbent provides you with enhanced retention of acidic compounds.
Specifications

Specifications

2.1 mm
6000 psi (415 Bar)
5 μm
1/Ea.
No
150 mm
Spherical
130 Å
L43
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers