Learn More
Waters Corporation XSelect CSH Fluoro-Phenyl Column
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases.
£877.50 - £1132.50
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L43 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
17133032
|
Waters Corporation
186005305 |
100 mm | 1 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17163032
|
Waters Corporation
186005310 |
50 mm | 2.1 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17173032
|
Waters Corporation
186005311 |
100 mm | 2.1 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17193042
|
Waters Corporation
186005331 |
150 mm | 2.1 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17103052
|
Waters Corporation
186005335 |
50 mm | 3 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17183042
|
Waters Corporation
186005330 |
100 mm | 2.1 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17183052
|
Waters Corporation
186005346 |
250 mm | 4.6 mm | 5 μm |
£1132.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17153042
|
Waters Corporation
186005323 |
75 mm | 4.6 mm | 3.5 μm |
£952.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17123032
|
Waters Corporation
186005304 |
50 mm | 1 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17133042
|
Waters Corporation
186005321 |
30 mm | 4.6 mm | 3.5 μm |
£907.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17163042
|
Waters Corporation
186005328 |
30 mm | 2.1 mm | 5 μm |
£877.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17143042
|
Waters Corporation
186005322 |
50 mm | 4.6 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17153052
|
Waters Corporation
186005343 |
75 mm | 4.6 mm | 5 μm |
£937.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17113052
|
Waters Corporation
186005336 |
100 mm | 3 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
10577290
|
Waters Corporation
186005324 |
100 mm | 4.6 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17133052
|
Waters Corporation
186005341 |
30 mm | 4.6 mm | 5 μm |
£877.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17153032
|
Waters Corporation
186005309 |
30 mm | 2.1 mm | 3.5 μm |
£907.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17103042
|
Waters Corporation
186005316 |
50 mm | 3 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17173042
|
Waters Corporation
186005329 |
50 mm | 2.1 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17123052
|
Waters Corporation
186005337 |
150 mm | 3 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17183032
|
Waters Corporation
186005312 |
150 mm | 2.1 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17113042
|
Waters Corporation
186005317 |
100 mm | 3 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17163052
|
Waters Corporation
186005344 |
100 mm | 4.6 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17133232
|
Waters Corporation
186005646 |
75 mm | 2.1 mm | 3.5 μm |
£952.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
17163232
|
Waters Corporation
186005649 |
75 mm | 3 mm | 3.5 μm |
£952.50
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
Description
- The non-endcapped sorbent provides you with enhanced retention of acidic compounds.
Specifications
6000 psi (415 Bar) | |
130 Å | |
L43 | |
Column |
Spherical | |
1/Ea. | |
No |