Learn More
Waters Corporation XSelect CSH Fluoro-Phenyl XP Column
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases.
Brand: Waters Corporation 186006122

Description
- The non-endcapped sorbent provides you with enhanced retention of acidic compounds.
- Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Specifications
4.6 mm | |
10000 psi (690 Bar) | |
2.5 μm | |
1/Ea. | |
No |
75 mm | |
Spherical | |
130 Å | |
L43 | |
eXtended Performance (XP) Column |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.