Learn More
Waters Corporation XTerra MS C8 Column
Encompass
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns that are designed to be compatible with mass spectrometry applications.
£885.00 - £1230.00
Specifications
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Pore Size 125 Å
Quantity 1/Ea.
USP Type L7
View More Specs
Products 26
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
13252560
Encompass
View Documents
Waters Corporation
186000401
50 mm 2.1 mm 3.5 μm
£954.00
Each
View alternative products
Save Up to 
»
13292590
Encompass
View Documents
Waters Corporation
186000441
150 mm 4.6 mm 3.5 μm
£1114.50
Each
View alternative products
Save Up to 
»
13222640
Encompass
View Documents
Waters Corporation
186000491
150 mm 4.6 mm 5 μm
£1086.00
Each
View alternative products
Save Up to 
»
13292740
Encompass
View Documents
Waters Corporation
186000603
50 mm 4.6 mm 2.5 μm
£982.50
Each
View alternative products
Save Up to 
»
13232560
Encompass
View Documents
Waters Corporation
186000399
30 mm 2.1 mm 3.5 μm
£925.50
Each
View alternative products
Save Up to 
»
13282560
Encompass
View Documents
Waters Corporation
186000405
100 mm 2.1 mm 3.5 μm
£1039.50
Each
View alternative products
Save Up to 
»
10083204
Encompass
View Documents
Waters Corporation
186000409
150 mm 2.1 mm 3.5 μm
£1114.50
Each
View alternative products
Save Up to 
»
13252570
Encompass
View Documents
Waters Corporation
186000413
30 mm 3 mm 3.5 μm
£925.50
Each
View alternative products
Save Up to 
»
13272570
Encompass
View Documents
Waters Corporation
186000415
50 mm 3 mm 3.5 μm
£954.00
Each
View alternative products
Save Up to 
»
10345054
Encompass
View Documents
Waters Corporation
186000419
100 mm 3 mm 3.5 μm
£1039.50
Each
View alternative products
Save Up to 
»
13232580
Encompass
View Documents
Waters Corporation
186000423
150 mm 3 mm 3.5 μm
£1114.50
Each
View alternative products
Save Up to 
»
13272580
Encompass
View Documents
Waters Corporation
186000427
100 mm 3.9 mm 3.5 μm
£1039.50
Each
View alternative products
Save Up to 
»
13222590
Encompass
View Documents
Waters Corporation
186000433
50 mm 4.6 mm 3.5 μm
£954.00
Each
View alternative products
Save Up to 
»
13262590
Encompass
View Documents
Waters Corporation
186000437
100 mm 4.6 mm 3.5 μm
£1039.50
Each
View alternative products
Save Up to 
»
13212600
Encompass
View Documents
Waters Corporation
186000447
50 mm 2.1 mm 5 μm
£925.50
Each
View alternative products
Save Up to 
»
13242600
Encompass
View Documents
Waters Corporation
186000451
100 mm 2.1 mm 5 μm
£994.50
Each
View alternative products
Save Up to 
»
13282600
Encompass
View Documents
Waters Corporation
186000455
150 mm 2.1 mm 5 μm
£1086.00
Each
View alternative products
Save Up to 
»
13252610
Encompass
View Documents
Waters Corporation
186000463
50 mm 3 mm 5 μm
£919.50
Each
View alternative products
Save Up to 
»
13292610
Encompass
View Documents
Waters Corporation
186000467
100 mm 3 mm 5 μm
£994.50
Each
View alternative products
Save Up to 
»
13232620
Encompass
View Documents
Waters Corporation
186000471
150 mm 3 mm 5 μm
£1086.00
Each
View alternative products
Save Up to 
»
13212630
Encompass
View Documents
Waters Corporation
186000479
150 mm 3.9 mm 5 μm
£1086.00
Each
View alternative products
Save Up to 
»
10017951
Encompass
View Documents
Waters Corporation
186000483
50 mm 4.6 mm 5 μm
£925.50
Each
View alternative products
Save Up to 
»
13282630
Encompass
View Documents
Waters Corporation
186000487
100 mm 4.6 mm 5 μm
£994.50
Each
View alternative products
Save Up to 
»
13242640
Encompass
View Documents
Waters Corporation
186000495
250 mm 4.6 mm 5 μm
£1195.50
Each
View alternative products
Save Up to 
»
13194439
Encompass
View Documents
Waters Corporation
186000879
30 mm 4.6 mm 5 μm
£885.00
Each
View alternative products
Save Up to 
»
13174649
Encompass
View Documents
Waters Corporation
186001471
250 mm 4.6 mm 3.5 μm
£1230.00
Each
View alternative products
Save Up to 
»
Description
Description
  • These first generation columns provide superior pH stability compared to a silica-based column
Specifications
Specifications
6000 psi (415 Bar)
125 Å
L7
Column
Spherical
1/Ea.
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XTerra MS C8 Column