Learn More
Waters Corporation XTerra Phenyl Column
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality.
£919.50 - £1230.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 125 Å |
Quantity | 1/Ea. |
USP Type | L11 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
13184519
![]() |
Waters Corporation
186001140 |
150 mm | 4.6 mm | 3.5 μm |
£1114.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13154559
![]() |
Waters Corporation
186001179 |
50 mm | 2.1 mm | 3.5 μm |
£976.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13114529
![]() |
Waters Corporation
186001143 |
150 mm | 3 mm | 3.5 μm |
£1114.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13194519
![]() |
Waters Corporation
186001141 |
50 mm | 3 mm | 3.5 μm |
£976.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13104529
![]() |
Waters Corporation
186001142 |
100 mm | 3 mm | 3.5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13124529
![]() |
Waters Corporation
186001144 |
50 mm | 4.6 mm | 5 μm |
£919.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13134529
![]() |
Waters Corporation
186001145 |
100 mm | 4.6 mm | 5 μm |
£994.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13144529
![]() |
Waters Corporation
186001146 |
150 mm | 4.6 mm | 5 μm |
£1086.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13174519
![]() |
Waters Corporation
186001139 |
100 mm | 4.6 mm | 3.5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13154529
![]() |
Waters Corporation
186001147 |
250 mm | 4.6 mm | 5 μm |
£1195.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13144559
![]() |
Waters Corporation
186001178 |
150 mm | 3.9 mm | 3.5 μm |
£1114.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13164559
![]() |
Waters Corporation
186001180 |
100 mm | 2.1 mm | 3.5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13174559
![]() |
Waters Corporation
186001181 |
150 mm | 2.1 mm | 3.5 μm |
£1114.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13194559
![]() |
Waters Corporation
186001184 |
150 mm | 3.9 mm | 5 μm |
£1086.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13104659
![]() |
Waters Corporation
186001474 |
250 mm | 4.6 mm | 3.5 μm |
£1230.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13134559
![]() |
Waters Corporation
186001177 |
100 mm | 3.9 mm | 3.5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- These first generation columns provide superior pH stability compared to a silica-based column
Specifications
6000 psi (415 Bar) | |
125 Å | |
L11 | |
Column |
Spherical | |
1/Ea. | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XTerra Phenyl Column