Learn More
Waters Corporation XTerra Phenyl Column
XTerra Phenyl columns are hybrid-based, reversed-phase columns that incorporate phenyl ligand functionality.
Brand: Waters Corporation 186001178

Description
- These first generation columns provide superior pH stability compared to a silica-based column
Specifications
3.9 mm | |
6000 psi (415 Bar) | |
3.5 μm | |
1/Ea. | |
Yes |
150 mm | |
Spherical | |
125 Å | |
L11 | |
Column |
Product Suggestions
Customers who viewed this item also viewed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.