Learn More
Macherey-Nagel™ Chromabond™ Lid and Gasket
For use with glass cabinet in SPE applications for 16 columns
Brand:  Macherey-Nagel™ 730176
 View more versions of this product
Product Code. 10194991
  • £516.00 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
Specifications
16 Columns
Lid and Gasket
Include: Luer Fittings and Valves
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Lid and Gasket > For Use With: For 16 Columns