Learn More
Macherey-Nagel™ Chromabond™ Lid and Gasket
For use with glass cabinet in SPE applications for 16 columns
Brand: Macherey-Nagel™ 730175
334.32 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.Specifications
12 Columns | |
Lid and Gasket | |
Luer Fittings and Valves | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Lid and Gasket > For Use With: For 12 Columns
Spot an opportunity for improvement?Share a Content Correction