missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ XV100 Series Safety Glasses
Highly economical eye protection with a wrap-around uni-lens for great peripheral vision and protection.
£4.05 - £5.05
Specifications
Certifications/Compliance (Europe) | EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 |
---|---|
For Use With (Application) | General use and protection, utilities, light and heavy manufacturing, biotech, ship building, construction, automotive manufacturing |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
Product Code | Brand | Frame Color | Lens Tint | LensType | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Frame Color | Lens Tint | LensType | Price | Quantity & Availability | ||||
12677443
|
Honeywell™
1028860 |
Clear | Clear |
£5.05
Each |
In Stock
Log in to see stock. ![]() |
|||||
15826655
|
Honeywell™
1029692 |
Gray | Gray |
£5.05
Each |
In Stock
Log in to see stock. |
|||||
15816655
|
Honeywell™
1029691 |
Yellow | Yellow |
£4.05
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
Description
- 10-base, wrap-around, uni-lens for great peripheral vision and protection
- Molded, universal nosepiece for easy fit
- Lightweight design and thin, shaped temples for all-day comfort
- Protects against impact (45 m/s)
- Well suited for use in light and heavy manufacturing, utilities, biotech, ship building, construction, and automotive manufacturing
Specifications
EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 | |
General use and protection, utilities, light and heavy manufacturing, biotech, ship building, construction, automotive manufacturing | |
Polycarbonate | |
Polycarbonate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ XV100 Series Safety Glasses