Learn More
Honeywell™ XV100 Series Safety Glasses
Click to view available options
Frame Color:
Clear
Gray
Yellow
Lens Tint:
Clear
Gray
Yellow
Description
- 10-base, wrap-around, uni-lens for great peripheral vision and protection
- Molded, universal nosepiece for easy fit
- Lightweight design and thin, shaped temples for all-day comfort
- Protects against impact (45 m/s)
- Well suited for use in light and heavy manufacturing, utilities, biotech, ship building, construction, and automotive manufacturing
Specifications
Specifications
Certifications/Compliance | EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 |
Lens Coating | Anti-fog |
Description | Model XV104 |
Certifications/Compliance (Europe) | EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 |
Lens Tint | Yellow |
For Use With (Application) | General use and protection, utilities, light and heavy manufacturing, biotech, ship building, construction, automotive manufacturing |
Frame Color | Yellow |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Honeywell™ XV100 Series Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction