Learn More
3M™ Over-the-Glass Visitor Safety Glasses Product Code.: 10655325
Add to basket
3M™ Over-the-Glass Visitor Safety Glasses
Product Code. 10655325
Clear
Click to view available options
Clear
Product Code. 10655325
Brand: 3M™ 7000062913
Edge
Add to basket
Add to basket
To be worn on their own or over current style prescription spectacles. 3M™ Over-the-Glass Visitor Glasses with integrated molded sideshields and browguards provide maximum protection.
Classic style visitor spectacles
  • Lightweight lenses
  • Moulded sideshields and browguards
  • Can be worn on their own or over current style prescription spectacles.
  • Integrated side-shields for maximum protection
  • Lens coatings provide scratch resistance
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Clear
Product Type Protective eyewear
For Use With (Application) Protection Against Low Energy Impact, UV Protection
UV Protection Yes
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ Over-the-Glass Visitor Safety Glasses >