missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ OX3000 Polycarbonate Cover Safety Glasses
Click to view available options
Color: Blue
Description
Classic polycarbonate visitors glasses with DX/UV
- Clear lenses with blue frame
- Economical price to performance ratio,
- Straight, flat, adjustable straps compatible with 3M™ Earmuffs
- Can be worn over prescription glasses
- Adjustable lens angle and temple length for optimal fit
- 5-stage adjustable temple length
- Soft and narrow strap ends
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Product Type | Safety Glasses; Compact |
For Use With (Application) | Construction, Forestry |
Frame Color | Blue, Black |
No. per Pack | 1 Ea. |
Temple Style | Soft and narrow |
Frame Material | Polycarbonate |
UV Protection | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction