missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylene, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Description
Xylene, Extra Pure, SLR

Specifications
Specifications
Melting Point | -34°C |
Color | Colorless |
Boiling Point | 136°C |
Quantity | 5 L |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Vapor Pressure | 8mbar at 20°C |
Viscosity | 0.6 mPaS at 20°C |
Formula Weight | 106.17g/mol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526020 | Certificate of Analysis | 10/02/2025 | X/0200/21,X/0200/PB17,X/0200/17,X/0200/08,X/0200/17Z,X/0200/25,X/0200/27 |
2526026 | Certificate of Analysis | 07/02/2025 | X/0200/21,X/0200/PB17,X/0200/17,X/0200/08,X/0200/17Z,X/0200/25,X/0200/27 |
2526025 | Certificate of Analysis | 07/02/2025 | X/0200/21,X/0200/PB17,X/0200/17,X/0200/08,X/0200/17Z,X/0200/25,X/0200/27 |
Safety and Handling



Product Identifier
- Xylene
- Danger
- Flammable liquid Category 3
- Aspiration toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Xylene, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction