missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange, sodium salt

£31.86 - £783.90

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10174740
View Documents
Thermo Scientific Chemicals
211220010
1 g Glass bottle
£31.86
1g
In Stock
to see stock.
Add to basket
 
10676802
View Documents Promotion Details
Thermo Scientific Chemicals
211220050
5 g Glass bottle
£90.05
5g
In Stock
to see stock.
Add to basket
 
10421831
View Documents
Thermo Scientific Chemicals
211220100
10 g Glass bottle
£159.80
10g
In Stock
to see stock.
Add to basket
 
11996931
View Documents
Thermo Scientific Chemicals
211220500
50 g Glass bottle
£783.90
50g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

3618-43-7
C31H28N2Na4O13S

Specifications

210.0°C
C31H28N2Na4O13S
Authentic
760.59
Pure
Complies
Crystalline Powder
3618-43-7
Solubility in water: 510g/L (20°C).
575 to 585nm (0.1N NaOH)
10.0% (110°C, 1 hr) max.
(for Zinc titration) Passes test
Brown to Brown/Red
Xylenol Orange, sodium salt
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.