missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus

Product Code. 15455459
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
500 mg
1 g
missing translation for 'unitSize'
1g
250mg
500mg
This item is not returnable. View return policy

Product Code. 15455459

missing translation for 'mfr': Thermo Scientific Chemicals J63195.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolyzes linear glucose polymers with beta-1,3-linkages | CAS: 37340-57-1

Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37340-57-1
Synonym Lyticase; Zymolyase 20T

Specifications

Color Off-white
Product Type Cell Lysis Enzyme
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Hydrolyzes linear glucose polymers with beta-1,3-linkages
Quantity 500 mg
compliance-icons
Product Identifier
  • Yeast Lytic Enzyme, Arthrobacter luteus
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.