Learn More
Checkers Yellow Jacket™ AMS™ 5 Channel Male Ramps
Yellow Jacket™ 5-channel AMS ramps can be combined with the Yellow Jacket™ 5-channel AMS center section to create a system that is fully expandable to hold an unlimited number of cables.
Brand:  Checkers PYJS0006BL
Product Code. 17792428
  • £113.00 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Yellow Jacket™ 5-channel AMS ramps can be combined with the Yellow Jacket™ 5-channel AMS center section to create a system that is fully expandable to hold an unlimited number of cables
  • By detaching the ramps, you can also quickly add cable runs and ADA-compatible side ramps to existing systems
  • The Yellow Jacket™ AMS™ readily handles heavy truck and bus traffic while providing easy access for pedestrians, carts, and wheelchairs
  • Hinged lids make for easy loading of cables and hoses
Specifications
Specifications
51 x 91 x 5 cm
Black
Polyethylene
5 Channel Ramps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Checkers Yellow Jacket™ AMS™ 5 Channel Male Ramps > Channel Ramp; Color: Black