Learn More
Gilson™ PIPETMAN™ M Pipettes
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
Brand: Gilson™ F81032
Description
- Motorized piston with adjustable piston speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
- Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
D10, DL10, DF10 or DFL10 tips | |
P12x300M | |
Pipetman | |
10 to 300 μL | |
1 Year |
Green | |
12 | |
Pipette | |
Ergonomic |
Product Suggestions
Customers who viewed this item also viewed.