missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Yellow Micro PTFE Stir Bar

Fisherbrand™ Yellow Micro PTFE Stir Bar is an inert and leak-free stirrer for a wide variety of applications where identification is important.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 16235851

  • £7.40 / Pack of 10
Estimated Shipment: 09-09-2024
to see stock.
Add to basket

Description

Description

  • Micro stirrer bars
  • For use where identification is of prime importance
  • PTFE coatings are not as inert as pure PTFE
  • Individually polished to stay cleaner for longer
Specifications

Specifications

3 mm
Yellow
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Micro
PTFE encapsulated Alnico V magnet core
10/Pk.
Yes
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers