missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11361799

Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11361799
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11361799

Brand: Thermo Scientific Alfa Aesar 012437.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for phosphors, ceramic capacitors, ferrite devices, pulsed lasers and catalysts. Use in improving the strength and mechanical properties of stainless steel. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

UN Number UN3178
Quantity 25 x 25 mm
Solubility Information It reacts slowly with water and is soluble in dilute acids.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.43g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Ytterbium foil, 0.1mm (0.004 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT21H007 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number012437.FI,012437.FF
Lot NumberZ05I002 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number012437.FI,012437.FF
Lot NumberB11X049 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012437.FI,012437.FF
Lot NumberD20W004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012437.FI,012437.FF
Lot NumberX17D045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012437.FI,012437.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.