missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11326228
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
19423-87-1
Cl3Yb
279.40
MFCD00149885
CKLHRQNQYIJFFX-UHFFFAOYSA-K
ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1
71317291
[Cl-].[Cl-].[Cl-].[Yb+3]
This item is not returnable. View return policy
19423-87-1
Cl3Yb
279.40
MFCD00149885
CKLHRQNQYIJFFX-UHFFFAOYSA-K
ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1
71317291
[Cl-].[Cl-].[Cl-].[Yb+3]

Ytterbium(III) chloride hydrate is used as an effective catalyst to prepare acetals by reacting with trimethyl orthoformate. It reacts with N-alkylsalicylideneimines to prepare yttrium(III) Schiff base complexes. It is widely used as NMR shift reagent, calcium ion probe and in DNA microarrays. Its anhydrous form acts as a Lewis acid and catalyzes the alkylation reactions such as aldol reaction and Pictet-Spengler reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150°C to 155°C
Color Green
Physical Form Wet Crystalline Aggregates
Quantity 10 g
Merck Index 14,10105
Solubility Information Soluble in water.
IUPAC Name ytterbium(3+);trichloride;hydrate
Formula Weight 279.40 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 2.58 g/mL
Assay (REO)
Chemical Name or Material Ytterbium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'