missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11356228

Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11356228
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11356228

Brand: Thermo Scientific Alfa Aesar 011194.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cl3H2OYb, CAS Number-19423-87-1, acmc-1bo6s, ytterbium 3+ chloride-water 1/3/1, ytterbium 3+ ion hydrate trichloride, ytterbium iii chloride hydrate, 50g, 99.99% (REO), White, 2.58, 233-800-5, 279.40 (Anhydrous), H315-H319-H335

Ytterbium(III) chloride hydrate is used as a catalyst in organic chemical synthesis. It is also used to track digestion in animals. The catalytic nature of this compound also has an application in DNA microarrays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-87-1
Molecular Formula Cl3Yb
Molecular Weight (g/mol) 279.40
MDL Number MFCD00149885
InChI Key CKLHRQNQYIJFFX-UHFFFAOYSA-K
Synonym ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1
PubChem CID 71317291
IUPAC Name ytterbium(3+);trichloride;hydrate
SMILES [Cl-].[Cl-].[Cl-].[Yb+3]

Specifications

Melting Point 150°C to 155°C
Color White
Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 279.40 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.58
Chemical Name or Material Ytterbium(III) chloride hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX24I037 Certificate TypeCertificate of Analysis Date07/11/2022 Catalog Number011194.18,011194.09
Lot NumberT18I018 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number011194.18,011194.09
Lot Number22800 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011194.18,011194.09
Lot NumberI06R029 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011194.18,011194.09
Lot Number22981 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011194.18,011194.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.